Assay Details
| Gene Symbol: fhl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004112.1 | NP_001004112.1 | 4 - 5 | 542 | 67 | |
| GenBank mRNA | BC079524.1 | - | 4 - 5 | 542 | 67 | |
More Information
Set Membership: |
|

fhl5
Genomic Map