Assay Details
| Gene Symbol: rrh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004654.1 | NP_001004654.1 | 5 - 6 | 773 | 77 | |
| GenBank mRNA | BC081524.1 | - | 5 - 6 | 773 | 77 | |
| BC164135.1 | - | 5 - 6 | 748 | 77 | ||
| KT008425.1 | - | 5 - 6 | 714 | 77 | ||
More Information
Set Membership: |
|

rrh
Genomic Map