Assay Details
Gene Symbol: txnl4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005953.1 | NP_001005953.1 | 3 - 4 | 435 | 77 | |
GenBank mRNA | BC083448.1 | - | 3 - 4 | 435 | 77 | |
BC164224.1 | - | 2 - 3 | 287 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
