Assay Details
| Gene Symbol: trpa1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007065.1 | NP_001007066.1 | 14 - 15 | 1674 | 91 | |
| XM_017358267.2 | XP_017213756.1 | 13 - 14 | 1669 | 91 | ||
| GenBank mRNA | AY677196.1 | - | 14 - 15 | 1674 | 91 | |
| EU826643.1 | - | 13 - 14 | 1643 | 91 | ||
More Information
Set Membership: |
|

trpa1a
Genomic Map