Assay Details
| Gene Symbol: trpa1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007065.1 | NP_001007066.1 | 17 - 18 | 1976 | 72 | |
| XM_017358267.2 | XP_017213756.1 | 16 - 17 | 1971 | 72 | ||
| GenBank mRNA | AY677196.1 | - | 17 - 18 | 1976 | 72 | |
| EU826643.1 | - | 16 - 17 | 1945 | 72 | ||
More Information
Set Membership: |
|

trpa1a
Genomic Map