Assay Details
Gene Symbol: trpa1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007065.1 | NP_001007066.1 | 23 - 24 | 2702 | 70 | |
XM_017358267.2 | XP_017213756.1 | 22 - 23 | 2697 | 70 | ||
GenBank mRNA | AY677196.1 | - | 23 - 24 | 2702 | 70 | |
EU826643.1 | - | 22 - 23 | 2671 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
