Assay Details
| Gene Symbol: cadm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007428.1 | NP_001007429.1 | 2 - 3 | 398 | 65 | |
| XM_005158111.4 | XP_005158168.1 | 2 - 3 | 914 | 65 | ||
| XM_005158112.2 | XP_005158169.1 | 2 - 3 | 422 | 65 | ||
| XM_005158113.4 | XP_005158170.1 | 2 - 3 | 908 | 65 | ||
| GenBank mRNA | BC085419.1 | - | 2 - 3 | 398 | 65 | |
| BC164711.1 | - | 2 - 3 | 248 | 65 | ||
| CU638749.1 | - | 2 - 3 | 398 | 65 | ||
More Information
Set Membership: |
|

cadm4
Genomic Map