Assay Details
| Gene Symbol: eif4g2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013443.2 | NP_001013461.2 | 19 - 20 | 2857 | 81 | |
| GenBank mRNA | AB096101.1 | - | 19 - 20 | 2705 | 81 | |
| BC109409.1 | - | 19 - 20 | 2721 | 81 | ||
| BC127398.1 | - | 19 - 20 | 2736 | 81 | ||
More Information
Set Membership: |
|

eif4g2b
Genomic Map