Assay Details
Gene Symbol: mtmr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014330.2 | NP_001014352.1 | 2 - 3 | 419 | 83 | |
GenBank mRNA | BC091956.1 | - | 2 - 3 | 423 | 83 | |
BC165859.1 | - | 2 - 3 | 186 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
