Assay Details
Gene Symbol: mtmr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014330.2 | NP_001014352.1 | 9 - 10 | 1110 | 69 | |
XM_005161411.4 | XP_005161468.1 | 8 - 9 | 1112 | 69 | ||
GenBank mRNA | BC091956.1 | - | 9 - 10 | 1114 | 69 | |
BC165859.1 | - | 9 - 10 | 877 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
