Assay Details
Gene Symbol: cbsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014345.2 | NP_001014367.2 | 14 - 15 | 1725 | 70 | |
GenBank mRNA | AY880251.1 | - | 14 - 15 | 1725 | 70 | |
BC091981.1 | - | 10 - 11 | 1066 | 70 | ||
BC164709.1 | - | 9 - 10 | 1062 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
