Assay Details
Gene Symbol: cbsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014345.2 | NP_001014367.2 | 3 - 4 | 496 | 79 | |
GenBank mRNA | AY880251.1 | - | 3 - 4 | 496 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
