Assay Details
Gene Symbol: cbsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014345.2 | NP_001014367.2 | 6 - 7 | 911 | 94 | |
GenBank mRNA | AY880251.1 | - | 6 - 7 | 911 | 94 | |
BC091981.1 | - | 2 - 3 | 252 | 94 | ||
BC164709.1 | - | 1 - 2 | 248 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
