Assay Details
Gene Symbol: cbsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014345.2 | NP_001014367.2 | 9 - 10 | 1215 | 68 | |
GenBank mRNA | AY880251.1 | - | 9 - 10 | 1215 | 68 | |
BC091981.1 | - | 5 - 6 | 556 | 68 | ||
BC164709.1 | - | 4 - 5 | 552 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
