Assay Details
Gene Symbol: erbb3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014826.1 | NP_001014826.1 | 12 - 13 | 1773 | 72 | |
XM_005162170.4 | XP_005162227.1 | 13 - 14 | 1873 | 72 | ||
XM_021469802.1 | XP_021325477.1 | 12 - 13 | 1865 | 72 | ||
GenBank mRNA | AY863052.1 | - | 12 - 13 | 1773 | 72 | |
BC162634.1 | - | 13 - 14 | 1663 | 72 |
Gene Symbol: LOC100149258 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128819.1 | NP_001122291.1 | 13 - 14 | 1663 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
