Assay Details
Gene Symbol: erbb3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014826.1 | NP_001014826.1 | 14 - 15 | 2006 | 95 | |
XM_005162170.4 | XP_005162227.1 | 15 - 16 | 2106 | 95 | ||
XM_021469802.1 | XP_021325477.1 | 14 - 15 | 2098 | 95 | ||
GenBank mRNA | AY863052.1 | - | 14 - 15 | 2006 | 95 | |
BC162634.1 | - | 15 - 16 | 1896 | 95 |
Gene Symbol: LOC100149258 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128819.1 | NP_001122291.1 | 15 - 16 | 1896 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
