Assay Details
Gene Symbol: erbb3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014826.1 | NP_001014826.1 | 19 - 20 | 2579 | 72 | |
XM_005162170.4 | XP_005162227.1 | 20 - 21 | 2679 | 72 | ||
XM_021469802.1 | XP_021325477.1 | 19 - 20 | 2671 | 72 | ||
GenBank mRNA | AY863052.1 | - | 19 - 20 | 2579 | 72 | |
BC162634.1 | - | 20 - 21 | 2469 | 72 |
Gene Symbol: LOC100149258 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128819.1 | NP_001122291.1 | 20 - 21 | 2469 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
