Assay Details
Gene Symbol: erbb3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014826.1 | NP_001014826.1 | 5 - 6 | 902 | 89 | |
XM_005162170.4 | XP_005162227.1 | 5 - 6 | 1002 | 89 | ||
XM_021469802.1 | XP_021325477.1 | 5 - 6 | 1003 | 89 | ||
GenBank mRNA | AY863052.1 | - | 5 - 6 | 902 | 89 | |
BC162634.1 | - | 5 - 6 | 792 | 89 |
Gene Symbol: LOC100149258 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128819.1 | NP_001122291.1 | 5 - 6 | 792 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
