Assay Details
Gene Symbol: erbb3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014826.1 | NP_001014826.1 | 9 - 10 | 1388 | 115 | |
XM_005162170.4 | XP_005162227.1 | 9 - 10 | 1488 | 115 | ||
XM_021469802.1 | XP_021325477.1 | 9 - 10 | 1489 | 115 | ||
GenBank mRNA | AY863052.1 | - | 9 - 10 | 1388 | 115 | |
BC162634.1 | - | 9 - 10 | 1278 | 115 |
Gene Symbol: LOC100149258 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128819.1 | NP_001122291.1 | 9 - 10 | 1278 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
