Assay Details
Gene Symbol: rspo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017358.1 | NP_001017358.1 | 5 - 6 | 1344 | 106 | |
XM_005158196.4 | XP_005158253.1 | 5 - 6 | 905 | 106 | ||
GenBank mRNA | AB035930.1 | - | 5 - 6 | 1344 | 106 | |
AB769408.1 | - | 5 - 6 | 762 | 106 | ||
BC139514.1 | - | 5 - 6 | 1157 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
