Assay Details
| Gene Symbol: mrpl27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020680.2 | NP_001018516.1 | 2 - 3 | 173 | 61 | |
| GenBank mRNA | BC095759.1 | - | 2 - 3 | 175 | 61 | |
More Information
Set Membership: |
|

mrpl27
Genomic Map