Assay Details
| Gene Symbol: ifng1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020793.1 | NP_001018629.1 | 2 - 3 | 212 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ifng1r| Gene Symbol: ifng1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020793.1 | NP_001018629.1 | 2 - 3 | 212 | 90 | |
Set Membership: |
|