Assay Details
Gene Symbol: ets2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001023580.1 | NP_001018874.1 | 3 - 4 | 315 | 59 | |
GenBank mRNA | BC095899.1 | - | 3 - 4 | 315 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
