Assay Details
Gene Symbol: stxbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025182.1 | NP_001020353.1 | 5 - 6 | 355 | 76 | |
XM_005161113.4 | XP_005161170.1 | 5 - 6 | 430 | 76 | ||
GenBank mRNA | BC097221.1 | - | 5 - 6 | 355 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
