Assay Details
| Gene Symbol: slc4a2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037237.1 | NP_001032314.1 | 10 - 11 | 1726 | 107 | |
| XM_005163511.4 | XP_005163568.1 | 12 - 13 | 1911 | 107 | ||
| XM_009298626.3 | XP_009296901.1 | 11 - 12 | 1886 | 107 | ||
| XM_009298629.3 | XP_009296904.1 | 10 - 11 | 1829 | 107 | ||
| XM_021480957.1 | XP_021336632.1 | 12 - 13 | 1851 | 107 | ||
| XM_021480959.1 | XP_021336634.1 | 11 - 12 | 1785 | 107 | ||
| GenBank mRNA | AY876015.1 | - | 10 - 11 | 1726 | 107 | |
More Information
Set Membership: |
|

slc4a2a
Genomic Map