Assay Details
| Gene Symbol: slc4a2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037237.1 | NP_001032314.1 | 17 - 18 | 2996 | 84 | |
| XM_005163511.4 | XP_005163568.1 | 19 - 20 | 3181 | 84 | ||
| XM_009298626.3 | XP_009296901.1 | 18 - 19 | 3156 | 84 | ||
| XM_009298629.3 | XP_009296904.1 | 17 - 18 | 3099 | 84 | ||
| XM_021480957.1 | XP_021336632.1 | 19 - 20 | 3121 | 84 | ||
| XM_021480959.1 | XP_021336634.1 | 18 - 19 | 3055 | 84 | ||
| GenBank mRNA | AY876015.1 | - | 17 - 18 | 2996 | 84 | |
| DQ005500.1 | - | 3 - 4 | 542 | 84 | ||
| DQ005501.1 | - | 2 - 3 | 338 | 84 | ||
More Information
Set Membership: |
|

slc4a2a
Genomic Map