Assay Details
Gene Symbol: prkchb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044806.1 | NP_001038271.1 | 10 - 11 | 1412 | 77 | |
XM_005160500.4 | XP_005160557.1 | 10 - 11 | 1637 | 77 | ||
GenBank mRNA | BC163148.1 | - | 10 - 11 | 1510 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
