Assay Details
Gene Symbol: prkchb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044806.1 | NP_001038271.1 | 1 - 2 | 340 | 86 | |
GenBank mRNA | BC163148.1 | - | 1 - 2 | 438 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
