Assay Details
Gene Symbol: prkchb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044806.1 | NP_001038271.1 | 3 - 4 | 552 | 67 | |
XM_005160500.4 | XP_005160557.1 | 3 - 4 | 777 | 67 | ||
GenBank mRNA | BC163148.1 | - | 3 - 4 | 650 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
