Assay Details
| Gene Symbol: mcat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045043.1 | NP_001038508.1 | 3 - 4 | 917 | 64 | |
| GenBank mRNA | BC133958.1 | - | 3 - 4 | 933 | 64 | |
More Information
Set Membership: |
|

mcat
Genomic Map