Assay Details
| Gene Symbol: furina | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045106.2 | NP_001038571.1 | 3 - 4 | 707 | 70 | |
| XM_009302953.3 | XP_009301228.1 | 3 - 4 | 501 | 70 | ||
| GenBank mRNA | BC139879.1 | - | 3 - 4 | 707 | 70 | |
| DQ499476.1 | - | 2 - 3 | 422 | 70 | ||
More Information
Set Membership: |
|

furina
Genomic Map