Assay Details
Gene Symbol: furina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045106.2 | NP_001038571.1 | 4 - 5 | 797 | 68 | |
XM_009302953.3 | XP_009301228.1 | 4 - 5 | 591 | 68 | ||
GenBank mRNA | BC139879.1 | - | 4 - 5 | 797 | 68 | |
DQ499476.1 | - | 3 - 4 | 512 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
