Assay Details
| Gene Symbol: furina | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045106.2 | NP_001038571.1 | 6 - 7 | 1008 | 64 | |
| XM_009302953.3 | XP_009301228.1 | 6 - 7 | 802 | 64 | ||
| GenBank mRNA | BC139879.1 | - | 6 - 7 | 1008 | 64 | |
| DQ499476.1 | - | 5 - 6 | 723 | 64 | ||
More Information
Set Membership: |
|

furina
Genomic Map