Assay Details
Gene Symbol: dgkaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045315.1 | NP_001038780.1 | 14 - 15 | 1385 | 93 | |
XM_005162328.4 | XP_005162385.1 | 15 - 16 | 1539 | 93 | ||
GenBank mRNA | BC117645.1 | - | 14 - 15 | 1385 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
