Assay Details
| Gene Symbol: dgkaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045315.1 | NP_001038780.1 | 18 - 19 | 1798 | 72 | |
| XM_005162328.4 | XP_005162385.1 | 19 - 20 | 1952 | 72 | ||
| GenBank mRNA | BC117645.1 | - | 18 - 19 | 1798 | 72 | |
More Information
Set Membership: |
|

dgkaa
Genomic Map