Assay Details
| Gene Symbol: exosc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045352.2 | NP_001038817.2 | 3 - 4 | 374 | 59 | |
| GenBank mRNA | BC122387.1 | - | 3 - 4 | 59 | ||
| BC164315.1 | - | 3 - 4 | 59 | |||
| EH452602.1 | - | 3 - 4 | 401 | 59 | ||
| EH561913.1 | - | 3 - 4 | 353 | 59 | ||
More Information
Set Membership: |
|

exosc5
Genomic Map