Assay Details
| Gene Symbol: neurl1ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076708.1 | NP_001070176.2 | 4 - 5 | 1335 | 74 | |
| GenBank mRNA | BC124268.1 | - | 4 - 5 | 1335 | 74 | |
| BC164216.1 | - | 3 - 4 | 1151 | 74 | ||
More Information
Set Membership: |
|

neurl1ab
Genomic Map