Assay Details
| Gene Symbol: pcsk5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021476634.1 | XP_021332309.1 | 13 - 14 | 1998 | 74 | |
| GenBank mRNA | DQ983232.1 | - | 13 - 14 | 74 | ||
More Information
Set Membership: |
|

pcsk5a
Genomic Map