Assay Details
| Gene Symbol: gas1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077801.1 | NP_001071269.1 | 1 | 2753 | 86 | |
| GenBank mRNA | BC125918.1 | - | 1 | 2753 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gas1b| Gene Symbol: gas1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077801.1 | NP_001071269.1 | 1 | 2753 | 86 | |
| GenBank mRNA | BC125918.1 | - | 1 | 2753 | 86 | |
Set Membership: |
|