Assay Details
| Gene Symbol: asphd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080045.1 | NP_001073514.1 | 3 - 4 | 1391 | 66 | |
| XM_005167384.4 | XP_005167441.1 | 3 - 4 | 1484 | 66 | ||
| GenBank mRNA | BC125876.1 | - | 3 - 4 | 1391 | 66 | |
More Information
Set Membership: |
|

asphd2
Genomic Map