Assay Details
Gene Symbol: itgb3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082948.1 | NP_001076417.1 | 7 - 8 | 1114 | 69 | |
GenBank mRNA | BC163207.1 | - | 7 - 8 | 1073 | 69 | |
DQ178646.1 | - | 7 - 8 | 1114 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
