Assay Details
Gene Symbol: med13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083838.1 | NP_001077307.1 | 10 - 11 | 2760 | 124 | |
GenBank mRNA | BC129032.1 | - | 10 - 11 | 2760 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
