Assay Details
| Gene Symbol: stra6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089448.1 | NP_001082917.1 | 5 - 6 | 567 | 92 | |
| XM_005159799.4 | XP_005159856.1 | 6 - 7 | 589 | 92 | ||
| GenBank mRNA | BC134162.1 | - | 5 - 6 | 567 | 92 | |
More Information
Set Membership: |
|

stra6l
Genomic Map