Assay Details
Gene Symbol: aga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110281.1 | NP_001103751.1 | 7 - 8 | 830 | 78 | |
XM_021481042.1 | XP_021336717.1 | 5 - 6 | 545 | 78 | ||
GenBank mRNA | BC154306.1 | - | 7 - 8 | 830 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
