Assay Details
| Gene Symbol: desma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328376.1 | NP_001315305.1 | 2 - 3 | 686 | 73 | |
| NM_130963.2 | NP_571038.2 | 2 - 3 | 686 | 73 | ||
| GenBank mRNA | EH433465.1 | - | 2 - 3 | 73 | ||
| U47113.2 | - | 2 - 3 | 657 | 73 | ||
More Information
Set Membership: |
|

desma
Genomic Map