Assay Details
| Gene Symbol: desma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328376.1 | NP_001315305.1 | 4 - 5 | 937 | 82 | |
| NM_130963.2 | NP_571038.2 | 4 - 5 | 937 | 82 | ||
| GenBank mRNA | BC092715.1 | - | 4 - 5 | 937 | 82 | |
| BC165607.1 | - | 4 - 5 | 907 | 82 | ||
| U47113.2 | - | 4 - 5 | 908 | 82 | ||
More Information
Set Membership: |
|

desma
Genomic Map