Assay Details
Gene Symbol: desma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328376.1 | NP_001315305.1 | 5 - 6 | 1066 | 78 | |
NM_130963.2 | NP_571038.2 | 5 - 6 | 1066 | 78 | ||
GenBank mRNA | BC092715.1 | - | 5 - 6 | 1066 | 78 | |
BC165607.1 | - | 5 - 6 | 1036 | 78 | ||
U47113.2 | - | 5 - 6 | 1037 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
