Assay Details
| Gene Symbol: desma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328376.1 | NP_001315305.1 | 6 - 7 | 1292 | 89 | |
| NM_130963.2 | NP_571038.2 | 6 - 7 | 1292 | 89 | ||
| GenBank mRNA | BC092715.1 | - | 6 - 7 | 1292 | 89 | |
| BC165607.1 | - | 6 - 7 | 1262 | 89 | ||
| EH459468.1 | - | 3 - 4 | 510 | 89 | ||
| U47113.2 | - | 6 - 7 | 1263 | 89 | ||
More Information
Set Membership: |
|

desma
Genomic Map