Assay Details
Gene Symbol: nipsnap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131033.2 | NP_571108.2 | 7 - 8 | 888 | 71 | |
GenBank mRNA | AJ249796.1 | - | 7 - 8 | 588 | 71 | |
BC122249.1 | - | 7 - 8 | 888 | 71 | ||
EH486911.1 | - | 3 - 4 | 609 | 71 | ||
EH513303.1 | - | 7 - 8 | 647 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
