Assay Details
Gene Symbol: cpeb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131427.2 | NP_571502.2 | 8 - 9 | 1231 | 86 | |
XM_021477394.1 | XP_021333069.1 | 8 - 9 | 1245 | 86 | ||
GenBank mRNA | AB011680.1 | - | 7 - 8 | 1195 | 86 | |
AF076918.1 | - | 8 - 9 | 1210 | 86 | ||
BC045467.1 | - | 8 - 9 | 1229 | 86 | ||
BC164038.1 | - | 8 - 9 | 1229 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
